missing translation for 'onlineSavingsMsg'
Learn More

SALM4/LRFN3 Antibody, Novus Biologicals™

Product Code. 18425121 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
25ul
missing translation for 'unitSize'
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
18425121 25ul 25µL
18064649 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 18425121 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP21419925ul

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

SALM4/LRFN3 Polyclonal specifically detects SALM4/LRFN3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Spécification

Antigen SALM4/LRFN3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias fibronectin type III, immunoglobulin and leucine rich repeat domains 1, FIGLER1, leucine rich repeat and fibronectin type III domain containing 3, leucine-rich repeat and fibronectin type-III domain-containing protein 3, MGC2656, SALM4, synaptic adhesion-like molecule 4, UNQ5865/PRO34192
Gene Symbols LRFN3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: EATAAVELTVGPPPPPQLANSTSCDPPRDGDPDALTPPSAAS
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79414
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Afficher plus Afficher moins

For Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.