missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ salm Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144457
This item is not returnable.
View return policy
Description
salm Polyclonal Antibody for Western Blot, ELISA
Required for the establishment of the posterior-most head and the anterior-most tail segments of the embryo. Probably function as a transcriptional regulator. Could repress the transcription of the tsh gene.
Specifications
| salm | |
| Polyclonal | |
| Unconjugated | |
| salm | |
| sal | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 34569 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.29 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P39770 | |
| salm | |
| Recombinant Drosophila melanogaster (Fruit fly) SALM protein. | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction