missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SALL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581471
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse SALL4.
Sall4 protein is encoded by the Sall4 gene located on chromosome 20 in humans and belongs to the Spalt-like protein family. It plays an important role in somatic stem cell maintenance and is studied as a stem cell biomarker. It is a zinc finger transcription factor that is upregulated during development and shown to interact with Oct4 and Sox2. It also forms a complex with Nanog to maintain the pluripotency of Embryonic Stem cells (ESCs). Several current research studies focus on its function as an oncogene and tumor biomarker. Some of the reported target genes of Sall4 include p53, Bmi-1, Bcl2 and XIAP.
Specifications
| SALL4 | |
| Polyclonal | |
| Unconjugated | |
| SALL4 | |
| 5730441M18Rik; AA407717; AL022809; AW536104; C330011P20Rik; C78083; C78563; dJ1112F19.1; DRRS; HSAL4; SALL4; sal-like 4; sal-like 4 (Drosophila); sal-like protein 4; spalt like transcription factor 4; spalt-like transcription factor 4; Tex20; Zinc finger protein 797; zinc finger protein SALL4; ZNF797 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 99377 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8BX22 | |
| SALL4 | |
| A synthetic peptide corresponding to the N-terminus of the Mouse SALL4. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction