missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SALL4 Antibody (CL12183) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21209V
This item is not returnable.
View return policy
Description
SALL4 Monoclonal antibody specifically detects SALL4 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SALL4 | |
| Monoclonal | |
| DyLight 405 | |
| dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797sal (Drosophila)-like 4 | |
| This antibody was developed using a synthetic peptide derived from Q9UJQ4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL12183 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 57167 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction