missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SALL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571337
This item is not returnable.
View return policy
Description
SALL2 Polyclonal Antibody for Western Blot
This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.
Specifications
| SALL2 | |
| Polyclonal | |
| Unconjugated | |
| SALL2 | |
| AI225809; AW559097; COLB; HSAL2; KIAA0360; mKIAA0360; Msal-2; p150(Sal2); SAL2; Sal-2; SALL2; sal-like 2; sal-like 2 (Drosophila); sal-like protein 2; spalt like transcription factor 2; spalt-like protein 2; spalt-like transcription factor 2; zinc finger protein 795; Zinc finger protein SALL2; zinc finger protein Spalt-2; ZNF795 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6297 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y467 | |
| SALL2 | |
| synthetic peptide directed towards the N terminal region of human SALL2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction