missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Saline Solution
Shop All Novacyt Group Products
Click to view available options
:
Saline Solution
Description
- Allows the rapid grouping of Streptococcus A , B, C, D, F & G.
Specifications
Specifications
| Chemical Name or Material | 150 mL Saline Solution Buffer |
| Boiling Point | Approx. 100°C |
| Color | Clear |
| pH | 7.4 |
| Physical Form | Liquid |
| Formulation | Sodium Chloride, Water |
| Packaging | Bottle |
| Grade | AnalaR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction