missing translation for 'onlineSavingsMsg'
Learn More

Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents

Product Code. 10183433
Change view
Click to view available options
Quantity:
1L
4L
20L
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10183433 4L 4 L
10785104 20L 20 L
10418353 1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183433 Supplier Fisher BioReagents Supplier No. BP13254
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Saline-Sodium Citrate
Concentration 2.9 to 3.1 M
pH 7
Physical Form Liquid
Quantity 4L
DNase DNase free
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.