missing translation for 'onlineSavingsMsg'
Learn More

Salicylsalicylic acid, 98%

Product Code. 11450597
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450597 100 g 100g
11440597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450597 Supplier Thermo Scientific Alfa Aesar Supplier No. B22444.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 552-94-3 | C14H10O5 | 258.229 g/mol

Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-94-3
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00020252
InChI Key WVYADZUPLLSGPU-UHFFFAOYSA-N
Synonym salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
PubChem CID 5161
ChEBI CHEBI:9014
IUPAC Name 2-(2-hydroxybenzoyl)oxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O

Specifications

Melting Point 132°C to 138°C
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes slowly in water.
Formula Weight 258.23
Percent Purity 98%
Chemical Name or Material Salicylsalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.