missing translation for 'onlineSavingsMsg'
Learn More

Salicylic acid, MedChemExpress

Product Code. p-200100204
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
16444267 50 g 50g
16438378 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 16444267

Brand: Medchem Express HYB0167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salicylic acid (2-Hydroxybenzoic acid) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.

Salicylic acid (2-Hydroxybenzoic acid) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.

Specifications

Chemical Name or Material Salicylic acid
CAS 69-72-7
Color White
Purity Grade Notes Research
Molecular Formula C7H6O3
Quantity 50 g
Synonym 2-Hydroxybenzoic acid
Solubility Information DMSO : 100 mg/mL (724.01 mM; Need ultrasonic) ∣H2O : 1 mg/mL (7.24 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1O
Molecular Weight (g/mol) 138.12
Formula Weight 138.12
Percent Purity 96.22%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.