missing translation for 'onlineSavingsMsg'
Learn More

Salicylic acid, MedChemExpress

Product Code. 16444267
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444267 50 g 50g
16438378 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16444267 Supplier Medchem Express Supplier No. HYB0167
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Salicylic acid (2-Hydroxybenzoic acid) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.

Salicylic acid (2-Hydroxybenzoic acid) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.

Specifications

Chemical Name or Material Salicylic acid
CAS 69-72-7
Color White
Purity Grade Notes Research
Molecular Formula C7H6O3
Quantity 50 g
Synonym 2-Hydroxybenzoic acid
Solubility Information DMSO : 100 mg/mL (724.01 mM; Need ultrasonic) ∣H2O : 1 mg/mL (7.24 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1O
Molecular Weight (g/mol) 138.12
Formula Weight 138.12
Percent Purity 96.22%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.