Learn More
Salicylic acid, 99%, Thermo Scientific Chemicals
£17.70 - £64.30
Chemical Identifiers
CAS | 69-72-7 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002439 |
InChI Key | YGSDEFSMJLZEOE-UHFFFAOYSA-N |
Synonym | salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone |
PubChem CID | 338 |
ChEBI | CHEBI:16914 |
IUPAC Name | 2-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493980
|
Thermo Scientific Alfa Aesar
A12253.30 |
250 g |
£17.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454953
|
Thermo Scientific Alfa Aesar
A12253.36 |
500 g |
£19.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464953
|
Thermo Scientific Alfa Aesar
A12253.0E |
2500 g |
£64.30
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Salicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSalicylic acid is used as a food preservative as well as in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in dye industry for making azo direct dye and for acidic mordant dyeing.
Solubility
Soluble in carbon terachloride, benzene, propanol, absolute ethanol, acetone, toluene and ether. Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases, iodine, iron and iron salts.
Chemical Identifiers
69-72-7 | |
138.12 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
338 | |
2-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002439 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
CHEBI:16914 | |
OC(=O)C1=CC=CC=C1O |
Specifications
69-72-7 | |
211°C (20mmHg) | |
MFCD00002439 | |
14,8332 | |
Soluble in carbon terachloride,benzene,propanol,absolute ethanol,acetone,toluene and ether. Slightly soluble in water. | |
OC(=O)C1=CC=CC=C1O | |
138.12 | |
CHEBI:16914 | |
99% | |
157°C (315°F) | |
Salicylic acid |
158°C to 160°C | |
C7H6O3 | |
774890 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
2-hydroxybenzoic acid | |
338 | |
138.12 | |
1.443 | |
Light sensitive |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c
H302-H318-H361d
missing translation for 'einecsNumber' : 200-712-3
missing translation for 'rtecsNumber' : VO0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Salicylic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Salicylic acid
RUO – Research Use Only