missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid 2-O-Beta-D-Glucoside, TRC

Product Code. 30006221
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006221 50 mg 50mg
30004928 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006221 Supplier Toronto Research Chemicals Supplier No. TRCS08815550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10366-91-3
Molecular Formula C13H16O8
Molecular Weight (g/mol) 300.26
Synonym 2-(β-D-Glucopyranosyloxy)-benzoic Acid, o-(Glucopyranosyloxy)-benzoic Acid, o-(β-D-Glucopyranosyloxy)-benzoic Acid, o-(β-D-Glucosyloxy)-benzoic Acid, 2-O-β-Glucopyranosylsalicylic Acid, Salicylic Acid 2-β-D-Glucoside, Salicylic Acid O-Glucoside
IUPAC Name 2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzoic acid
SMILES OC[C@H]([C@@H](O)[C@H](O)[C@H]1O)O[C@H]1OC2=CC=CC=C2C(O)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H16O8/c14-5-8-9(15)10(16)11(17)13(21-8)20-7-4-2-1-3-6(7)12(18)19/h1-4,8-11,13-17H,5H2,(H,18,19)/t8-,9-,10+,11-,13-/m1/s1
Formula Weight 300.08
Percent Purity >95%
Chemical Name or Material Salicylic Acid 2-O-Beta-D-Glucoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.