missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, ≥99%, MP Biomedicals™

Product Code. 11476222 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476222 25 g 25g
11486222 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476222 Supplier MP Biomedicals Supplier No. 0210283525
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O

Specifications

Color White
Quantity 25 g
Formula Weight 137.1
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Salicylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.