missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, ≥99.0% (T), Honeywell™ Fluka™

Product Code. 15607290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Poly bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607290 1 g Poly bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15607290 Supplier Honeywell-Fluka Supplier No. 842281G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 65-45-2
Melting Point 140°C to 144°C
Packaging Poly bottle
Quantity 1 g
Molecular Formula C7H7NO2
Linear Formula 2-(HO)C6H4CONH2
Physical Form Solid
MDL Number MFCD00007978
UN Number NONH for all modes of transport
Beilstein 742439
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=CC=C1O
IUPAC Name 2-hydroxybenzamide
Molecular Weight (g/mol) 137.14
PubChem CID 5147
ChEBI CHEBI:32114
Formula Weight 137.14g/mol
Percent Purity ≥99.0% (T)
Chemical Name or Material Salicylamide, Matrix substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.