missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldoxime, TRC

Product Code. 30427981
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427981 500 mg 500mg
30413405 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427981 Supplier Toronto Research Chemicals Supplier No. TRCS881485500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylaldoxime
CAS 94-67-7
Molecular Formula C7H7NO2
Quantity 500 mg
InChI Formula InChI=1S/C7H7NO2/c9-7-4-2-1-3-6(7)5-8-10/h1-5,9-10H
SMILES ON=Cc1ccccc1O
IUPAC Name 2-(hydroxyiminomethyl)phenol
Molecular Weight (g/mol) 137.13
Formula Weight 137.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.