missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldoxime, 98%

Product Code. 10216790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216790 100 g Glass bottle 100g
10062790 25 g Glass bottle 25g
10056143 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216790 Supplier Thermo Scientific Acros Supplier No. 207401000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-67-7 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 94-67-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00002120
InChI Key GFCNBJDXQMZOOC-WAYWQWQTSA-N
Synonym salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol
PubChem CID 6740756
IUPAC Name 6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES ON\C=C1\C=CC=CC1=O

Tekniske data

Melting Point 56°C to 59°C
Color Bordeaux to Purple-Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CH=NOH
Quantity 100 g
Beilstein 08, 49
Merck Index 15, 8464
Formula Weight 137.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Salicylaldoxime
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Salicylaldoxime
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.