missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, TRC

Product Code. 30138342
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138342 500 g 500g
30133812 100 g 100g
30040448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138342 Supplier Toronto Research Chemicals Supplier No. TRCS088145500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7 H6 O2
Molecular Weight (g/mol) 122.12
Synonym Salicylaldehyde, Benzaldehyde, 2-hydroxy-, Salicylaldehyde (8CI), 2-Hydroxybenzaldehyde, 2-Formylphenol, 2-Hydroxy-1-benzaldehyde, NSC 112278, NSC 49178, NSC 83559, NSC 83560, NSC 83561, NSC 83562, NSC 97202, Salicylal, Salicylic aldehyde, o-Formylphenol, o-Hydroxybenzaldehyde
IUPAC Name 2-hydroxybenzaldehyde
SMILES Oc1ccccc1C=O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H
Formula Weight 122.0368
Percent Purity >95%
Chemical Name or Material Salicylaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.