missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde Azine, Mikromol™

Product Code. 17940886
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940886 100 mg 100mg
17903686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17940886 Supplier Mikromol Supplier No. MM0334.08
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Salicylaldehyde Azine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylaldehyde Azine
CAS 959-36-4
Analyte or Component Names Salicylaldehyde Azine
Shipping Condition Room Temperature
Molecular Formula C14 H12 N2 O2
Certifications/Compliance ISO 17025
Synonym Salicylaldehyde Azine, Benzaldehyde, 2-hydroxy-, [(2-hydroxyphenyl)methylene]hydrazone (9CI), 2,2'-Dihydroxybenzalazine, Bis[2-hydroxybenzaldehyde] hydrazone, N,N'-Bis(salicylidene)hydrazine, NSC 695048, NSC 864, Salazine, Salicylalazine, Salicylaldazine, Salicylazine, o-Hydroxybenzaldazine
InChI Formula InChI=1S/C14H12N2O2/c17-13-7-3-1-5-11(13)9-15-16-10-12-6-2-4-8-14(12)18/h1-10,17-18H/b15-9+,16-10+
SMILES Oc1ccccc1\C=N\N=C\c2ccccc2O
IUPAC Name 2-[(E)-[(E)-(2-hydroxyphenyl)methylidenehydrazinylidene]methyl]phenol
Molecular Weight (g/mol) 240.26
Quantity 100 mg
Formula Weight 240.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.