missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde azine, 97%

Product Code. 11498793
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498793 10 g 10g
11408803 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498793 Supplier Thermo Scientific Alfa Aesar Supplier No. A10527.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 959-36-4 | C14H12N2O2 | 240.262 g/mol

Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylaldehyde Azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.

Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-36-4
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.262
MDL Number MFCD00043496
InChI Key SPEXYYIULCBQJR-UHFFFAOYSA-N
Synonym salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone
PubChem CID 6849893
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1

Specifications

Melting Point 216°C to 222°C
Assay Percent Range 97%
Quantity 10 g
Beilstein 1913597
Solubility Information Soluble in dimethyl sulfoxide,methanol and ethyl acetate. Slightly soluble in water.
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material Salicylaldehyde Azine
compliance-icons
Product Identifier
  • Salicylaldehyde azine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.