missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%

Product Code. 11474924
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474924 2500 g 2500g
11454924 100 g 100g
11464924 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474924 Supplier Thermo Scientific Alfa Aesar Supplier No. A13833.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.

Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point -7°C
Density 1.168
Boiling Point 197°C
Flash Point 76°C (168°F)
Assay Percent Range 99%
Odor Almond-like
Refractive Index 1.573
Quantity 2500 g
Beilstein 471388
Sensitivity Air and light sensitive
Merck Index 14,8326
Solubility Information Miscible with alcohol,ether,acetone,benzene and most organic solvents. Slightly miscible with water.
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.