missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldazine, MP Biomedicals

Product Code. 11402776 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402776 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402776 Supplier MP Biomedicals Supplier No. 0521809080
  • ONLINE EXCLUSIVE PRICE
    £3280.00
    £3074.40 / 100g
    Save £205.60
    6% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Salicylaldazine, MP Biomedicals, Quantity: 100g, Molecular Weight: 240.262, CAS: 959-36-4, InChI Key: SPEXYYIULCBQJR-UHFFFAOYSA-N, IUPAC Name: 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one, Molecular Formula: C14H12N2O2, PubChem CID: 6849893

Specifications

Chemical Name or Material Salicylaldazine
CAS 959-36-4
Molecular Formula C14H12N2O2
Quantity 100 g
Synonym salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone
InChI Key SPEXYYIULCBQJR-UHFFFAOYSA-N
SMILES C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 240.262
PubChem CID 6849893
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.