missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldazine, 99%

Product Code. 15515226 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15515226 5 g 5g
15377995 25 g 25g
15525226 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15515226 Supplier Thermo Scientific Acros Supplier No. 456940050
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 959-36-4 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-36-4
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
InChI Key SPEXYYIULCBQJR-UHFFFAOYSA-N
Synonym salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone
PubChem CID 6849893
IUPAC Name 6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1

Specifications

Melting Point 218.0°C to 220.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 240.26
Percent Purity 99%
Chemical Name or Material Salicylaldazine
compliance-icons
Product Identifier
  • Salicylaldehyde azine, 99%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.