missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SALF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549902
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SALF protein.
Endocytosis of cell surface proteins is mediated by a complex molecular machinery that assembles on the inner surface of the plasma membrane. This gene encodes one of two human homologs of the Drosophila melanogaster stoned B protein. This protein is related to components of the endocytic machinery and exhibits a modular structure consisting of an N-terminal proline-rich domain, a central region of homology specific to the human stoned B-like proteins, and a C-terminal region homologous to the mu subunits of adaptor protein (AP) complexes. Co-transcription of this gene and the neighboring downstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
Specifications
| SALF | |
| Polyclonal | |
| Unconjugated | |
| STON1 | |
| 4921524J06Rik; C86231; Salf; Sblf; STN1; STNB1; STON1; STON1-GTF2A1L; STON1-GTF2A1L protein; STON1-GTF2A1L readthrough; stoned B homolog 1; stoned B/TFIIA-alpha/beta-like factor; stoned B-like factor; stoned-b1; stonin 1; Stonin1; stonin-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11037 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y6Q2 | |
| STON1 | |
| Synthesized peptide derived from C-terminal of human STON1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction