missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Salbutamol hemisulfate
GREENER_CHOICE

Product Code. 30001413
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001413 50 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001413 Supplier Tocris Bioscience™ Supplier No. 0634/250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β2-adrenoceptor agonist

Salbutamol hemisulfate is a selective β2-adrenergic agonist with 29-fold higher selectivity for β2 over β1 receptors. Salbutamol elevates fibroblast growth factor 20 (FGF20) in nigrostriatal neurons, and is neuroprotective in a mouse model of Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

CAS 51022-70-9
Quantity 50 mg
Molecular Formula C13H23NO7S
Formula Weight 576.7
Purity >99%
Molecular Weight (g/mol) 337.387
Chemical Name or Material Salbutamol hemisulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.