missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol hemisulfate, MedChemExpress

Product Code. 16442136
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442136

Brand: Medchem Express HYB0436

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting β2 adrenergic receptor agonist Target: β2 Adrenergic Receptor Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting, selective beta2-adrenergic receptor agonist used in the treatment of asthma and COPD.

Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting β2 adrenergic receptor agonist

Specifications

Chemical Name or Material Salbutamol hemisulfate
CAS 51022-70-9
Color White
Purity Grade Notes Research
Molecular Formula C13H22NO5S0.5
Quantity 500 mg
Synonym Albuterol hemisulfateAH-3365 hemisulfate
Solubility Information H2O : 100 mg/mL (347.05 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (6.94 mM; Need ultrasonic)
SMILES OC(C1=CC(CO)=C(O)C=C1)CNC(C)(C)C.[0.5H2SO4]
Molecular Weight (g/mol) 288.14
Formula Weight 288.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.