missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol hemisulfate, 98+%, Thermo Scientific™

£29.80 - £111.00

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C13H23NO7S
Molecular Weight (g/mol) 337.387
MDL Number MFCD00055200
InChI Key OVICLFZZVQVVFT-UHFFFAOYSA-N
Synonym salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
PubChem CID 9884233
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
SMILES CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478467
SDS View Documents
Thermo Scientific Alfa Aesar
A18544.03
1g
£29.80
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11488467
SDS View Documents
Thermo Scientific Alfa Aesar
A18544.06
5g
£111.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

Solubility
Soluble in water and 1M sodium hydroxide (50 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

51022-70-9
337.387
OVICLFZZVQVVFT-UHFFFAOYSA-N
9884233
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O
C13H23NO7S
MFCD00055200
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid

Specifications

51022-70-9
MFCD00055200
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
OVICLFZZVQVVFT-UHFFFAOYSA-N
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
9884233
99%
C13H23NO7S
Light sensitive
Soluble in water and 1M sodium hydroxide (50mg/ml).
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O
337.387
576.7
Salbutamol sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H361fd

missing translation for 'einecsNumber' : 256-916-8

missing translation for 'rtecsNumber' : CZ6430100

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Salbutamol sulphate

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contain: Bis[(tert-butyl)(.beta.,3,4-trihydroxyphenethyl)ammonium] sulphate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.