missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Sal 003

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Sal 003 is a cell-permeable inhibitor of cellular phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF2α). Analog of salubrinal (Cat. No. 2347) with improved aqueous solubility. Shown to prevent the induction of hippocampal long-term potentiation (LTP) and memory formation (LTM) in mice.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Sal 003 |
| Purity | >99% |
| Molecular Weight (g/mol) | 463.21 |
| Inhibitors | Sal 003 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction