missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001

Product Code. 11700418
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
Diameter (English) Outer:
1 in.
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
7/16 in.
7/8 in.
Thickness (English):
1/16 in.
1/8 in.
Unit Size:
15m
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer Thickness (English) unitSize
11700418 5/16 in. 7/16 in. 1/16 in. 15m
11780408 3/16 in. 5/16 in. 1/16 in. 15m
11790408 1/4 in. 3/8 in. 1/16 in. 15m
11760408 1/16 in. 3/16 in. 1/16 in. 15m
11770408 1/8 in. 1/4 in. 1/16 in. 15m
11710418 3/8 in. 1/2 in. 1/16 in. 15m
11720418 1/2 in. 3/4 in. 1/8 in. 15m
11730418 5/8 in. 7/8 in. 1/8 in. 15m
11740418 3/4 in. 1 in. 1/8 in. 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11700418 Leverandør Saint-Gobain Leverandørnr. 9643008

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications

Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.

  • Does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Crystal clear for easy visual flow monitoring
  • Meets FDA criteria for food contact
  • Will not degrade over time
  • Will not release hazardous byproducts when properly incinerated
  • Does not yield any taste
  • Helps ensure accurate results from analytical instrument tests

Ordreinfo

All products sold in 50-foot (15.24m) lengths

Overholdelse

Compliant with FDA 21 CFR, 177.2600

†At 73°F (23°C).

Tekniske data

Certifications/Compliance Compliant with FDA 21 CFR, 177.2600
Diameter (English) Inner 5/16 in.
Diameter (Metric) Inner 7.9 mm
Diameter (English) Outer 7/16 in.
Diameter (Metric) Outer 11.1 mm
Overall Length (English) 50 ft.
Thickness (English) 1/16 in.
Thickness (Metric) 1.6 mm
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.