missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
Diameter (English) Outer:
1 in.
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
7/16 in.
7/8 in.
Thickness (English):
1/16 in.
1/8 in.
Unit Size:
15m
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
- Does not contaminate fluids
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
- Crystal clear for easy visual flow monitoring
- Meets FDA criteria for food contact
- Will not degrade over time
- Will not release hazardous byproducts when properly incinerated
- Does not yield any taste
- Helps ensure accurate results from analytical instrument tests
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
†At 73°F (23°C).
Specifications
Specifications
| Certifications/Compliance | Compliant with FDA 21 CFR, 177.2600 |
| Diameter (English) Inner | 5/8 in. |
| Diameter (Metric) Inner | 15.9 mm |
| Diameter (English) Outer | 7/8 in. |
| Diameter (Metric) Outer | 22.2 mm |
| Overall Length (English) | 50 ft. |
| Thickness (English) | 1/8 in. |
| Thickness (Metric) | 3.2 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction