missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
- Does not contaminate fluids
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
- Crystal clear for easy visual flow monitoring
- Meets FDA criteria for food contact
- Will not degrade over time
- Will not release hazardous byproducts when properly incinerated
- Does not yield any taste
- Helps ensure accurate results from analytical instrument tests
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
†At 73°F (23°C).
Specifications
Specifications
| Certifications/Compliance | Compliant with FDA 21 CFR, 177.2600 |
| Diameter (English) Inner | 1/2 in. |
| Diameter (Metric) Inner | 12.7 mm |
| Diameter (English) Outer | 3/4 in. |
| Diameter (Metric) Outer | 19.1 mm |
| Overall Length (English) | 50 ft. |
| Thickness (English) | 1/8 in. |
| Thickness (Metric) | 3.2 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?