missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Chemfluor™ Fluoropolymer Tubing

Product Code. 11735179
Click to view available options
Diameter (English) Inner:
0.063 in.
0.13 in.
Diameter (English) Outer:
0.13 in.
1/4 in.
Material:
Fluoropolymer
PTFE
Unit Size:
300m
Each

Product Code. 11735179

Brand: Saint-Gobain 9600002

Only null left
Add to basket
Add to basket

Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals

  • Contains no additives or plasticizers
  • Inert to virtually all chemicals
  • Nonprotein binding

Order Info

In packs of 50 ft. (15m)

Compliance

Complies with FDA criteria

Specifications

Certifications/Compliance Complies with FDA criteria
Diameter (English) Inner 0.063 in.
Diameter (Metric) Inner 1.6 mm
Material PTFE
Diameter (English) Outer 0.13 in.
Diameter (Metric) Outer 3.2 mm
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 mm
For Use With (Application) Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.