missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SAHARA Acrylic S19T Heated Bath Circulators

Ideal when visibility of your application is required, with efficient heating from ambient plus 13°C to a maximum of 80°C.

£1444.88 - £2250.57


Products 4
Product Code Brand Control Voltage Hertz Price Quantity & Availability  
Product Code Brand Control Voltage Hertz Price Quantity & Availability  
11600901 Thermo Scientific™
155-2191
AC150 230V 50Hz £1999.91
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
11610921 Thermo Scientific™
156-2191
AC200 230V 50Hz £2250.57
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
11640861 Thermo Scientific™
153-2191
SC150 230V 50Hz £1801.17
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
11650841 Thermo Scientific™
152-2191
SC100 230V 50Hz £1444.88
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
Description

Description

Environment-friendly Design: Save up to 80% on energy costs and thousands of kilowatt hours during the life of the system. All units are RoHS-compliant (lead- and mercury-free) and use recycled packaging.

Safe Operation: All units are UL/CSA compliant, assuring safe operation in the unlikely event of a fault condition. 115/60 Hz units are UL listed or recognized, and all units are UL/CSA and RoHS compliant. IQ/OQ compliance is optional.

Ease of Use: All thermostats feature an intuitive user interface with bright display to view critical readings. Each system comes with a quick-start guide for simple setup and operation. Thermostat can be turned and positioned on all four sides on the bath for easy viewing.

Superior Warranty and Service: All solutions come with a 36-month warranty and are backed by Thermo Scientific service and support worldwide.

  • Capacity: 19L

  • Thermostats are attached by a sturdy bath bridge, can be positioned 90° on all four sides of the bath for easy viewing
  • Transparent acrylic reservoir is ideal for applications requiring good visibility
  • Temperatures are maintained from ambient plus 13° to 80°C

SC 100 controller

  • Features 3 languages, acoustic and optical alarms, and five user-settable programs
  • Two pump speeds allow adjustement of water flow within bath to reduce turbulence

SC 150 controller

  • Additional features include a USB port, a low-liquid level alarm, and an automatic controller shut-down at detection of excessive high temperature, low liquid level, or motor overload

AC 150 controller

  • Powerful force suction pump

AC 200 controller

  • Additional features include a ramp programming with 10 steps, a Real Time clock, and a USB port

Includes:

Communication cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation, 6ft. power cord

Recommended for:

  • Calibration
  • Bioreactors
  • Rotary Evaporators
  • Condensers
  • Sample/Material Testing
  • Sample/Material Preparation
  • Warranty varies by country, please contact us for details