missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SAHA
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
- SAHA (suberoylanilide hydroxamic acid) is a potent inhibitor of class I and class II histone deacetylases (HDACs)
- This compound inhibits the activities of HDAC1 and HDAC3 (IC50 = 10 nM and 20 nM, respectively; Richon et al.) SAHA exhibits antiproliferative properties and can cause cell cycle arrest at G1 (Marks & Breslow)
- Improves reprogramming efficiency from human and mouse somatic to induced pluripotent stem cells (Huangfu et al.)
- Downregulates homologous recombination and leads to apoptosis in ovarian cancer cells when used in combination with olaparib (Konstantinopoulos et al.)
- SAHA downregulates Nanog expression, which leads to proliferation inhibition in head and neck cancer stem cells (Kumar et al.)
Specifications
Specifications
| CAS | 149647-78-9 |
| Synonym | Suberoylanilide hydroxamic acid; Vorinostat; Zolinza™ |
| Quantity | 500 mg |
| Molecular Formula | C14H20N2O3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction