missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress SAH

Product Code. 16477788
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477788 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477788 Supplier MedChemExpress Supplier No. HY19528
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SAH (S-Adenosylhomocysteine) is an amino acid derivative and a modulartor in several metabolic pathways. It is an intermediate in the synthesis of cysteine and adenosine. SAH is an inhibitor for METTL3-METTL14 heterodimer complex (METTL3-14) with an IC50 of 0.9 μM.

Specifications

Molecular Weight (g/mol) 384.41 g/mol
Product Type Natural Products
Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 97.7%
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (325.17 mM∣Need ultrasonic)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 979-92-0
Synonym SAH (S-Adenosylhomocysteine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.