missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAG Mouse anti-Bovine, Human, Clone: S128, Abnova™
Mouse monoclonal antibody raised against partial recombinant bovine SAG.
Brand: Abnova MAB12504.50uL
This item is not returnable.
View return policy
Specifications
| SAG | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:5000) The optimal working dilution should be determined by the end user. | |
| SAG | |
| Recombinant protein corresponding to the C-terminus region of bovine SAG. | |
| 50 μL | |
| Primary | |
| Bovine, Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| S128 | |
| Mouse monoclonal antibody raised against partial recombinant bovine SAG. | |
| In PBS (10mM sodium azide) | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 280922 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction