missing translation for 'onlineSavingsMsg'
Learn More

SAG, MedChemExpress

Product Code. 16488107
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488107 5 mg 5mg
16435398 10 mg 10mg
16411587 50 mg 50mg
16403336 25 mg 25mg
16081282 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16488107 Supplier Medchem Express Supplier No. HY128485mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SAG is a potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM). SAG activates the Hedgehog signaling pathway and counteracts Cyclopamine (HY-17024) inhibition of Smo.

SAG is a potent Smoothened (Smo) receptor agonist (EC50=3 nM

Specifications

Chemical Name or Material SAG
CAS 912545-86-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H28ClN3OS
Quantity 5 mg
Solubility Information DMSO : ≥ 38 mg/mL (77.54 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=C(Cl)C2=CC=CC=C2S1)N([C@H]3CC[C@H](NC)CC3)CC4=CC=CC(C5=CC=NC=C5)=C4
Molecular Weight (g/mol) 490.06
Formula Weight 490.06
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.