missing translation for 'onlineSavingsMsg'
Learn More

SAG hydrochloride, MedChemExpress

Product Code. 16441309
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16441309

Brand: Medchem Express HY12848B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAG hydrochloride is a potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM). SAG hydrochloride activates the Hedgehog signaling pathway and counteracts Cyclopamine (HY-17024) inhibition of Smo.

SAG hydrochloride is a potent Smoothened (Smo) receptor agonist (EC50=3 nM

Specifications

Chemical Name or Material SAG hydrochloride
CAS 2095432-58-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H29Cl2N3OS
Quantity 1 mg
Solubility Information DMSO : 21.67 mg/mL (41.16 mM; Need ultrasonic)
SMILES O=C(C1=C(Cl)C2=CC=CC=C2S1)N([C@H]3CC[C@H](NC)CC3)CC4=CC=CC(C5=CC=NC=C5)=C4.Cl
Molecular Weight (g/mol) 526.52
Formula Weight 526.52
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.