missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SAG dihydrochloride

Product Code. 18783999
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18783999 1 mg 1mg
18053842 10 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18783999 Supplier R&D Systems™ Supplier No. 6390/1
  • ONLINE EXCLUSIVE PRICE
    £226.20
    £196.35 / 1mg
    Save £29.85
    13% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dihydrochloride salt of SAG; Smo receptor agonist

SAG dihydrochloride is a water-soluble dihydrochloride salt of SAG (Cat. No. 4366). SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and induces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide dihydrochloride
CAS 2702366-44-5
Assay Percent Range >99%
Quantity 1 mg
Product Line Tocris
Molecular Formula C28H28ClN3OS.2HCl
Formula Weight Observed MW: 562.98
Purity 98%
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.