missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SAG
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- SAG (Smoothened Agonist) is a chlorobenzothiophene-containing compound which acts as an activator of the G protein-coupled receptor Smoothened (SMO, EC50 = 3 nM; Chen et al.) SMO is a component of the Hedgehog signaling pathway, which is translocated to the primary cilium after stimulation of the Patched receptor by Hedgehog family ligands, leading to pathway activation
- SAG activates SMO via direct binding to the heptahelical bundle (Kd= 59 nM), stabilizing a specific conformation of SMO in cilia and leading to increased downstream gene expression (Rohatgi et al.) SAG abrogates cyclopamine inhibition of SMO, indicating that it acts downstream of cyclopamine (Frank-Kamenetsky et al.; Chen et al.; Lewis & Krieg)
Specifications
Specifications
| CAS | 912545-86-9 |
| Synonym | Smoothened Agonist |
| Quantity | 10 mg |
| Molecular Formula | C28H28ClN3OS |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction