Learn More
Pro-Lab Diagnostics™ Safranin Stain
Use for Gram staining of bacteria. Pro-Lab Diagnostics™ Safranin Stain is designed to be ready to use.
Brand: Pro-Lab Diagnostics™ PL7013
Code : 04
Additional Details : CAS Number : 64-17-5 Weight : 1.00000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions in a selection of convenient and economical sizes.
- Manufactured in accordance with all current regulatory requirements
- CE labelled
- Strict quality control and filtering techniques prior to release
- Also available are a range of convenient staining kits for Gram, TB and Cryptosporidium staining
Specifications
1 L | |
CE marked (98/78/EC) | |
Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria | |
Ethanol 10 - <25% , Methanol 1 - <2.5% , Glycerol 1 - <2.5% | |
Gram |
Safranin | |
Liquid | |
Dark Red | |
Keep at temperature not exceeding 20°C |
Safety and Handling
- Safranin
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash contaminated skin thoroughly after handling.
- P370+P378-In case of fire: Use foam, carbon dioxide, dry powder or water fog to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol