missing translation for 'onlineSavingsMsg'
Learn More

SAFit2, MedChemExpress

Product Code. 16466447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466447 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16466447 Supplier Medchem Express Supplier No. HY102080
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SAFit2 is a highly potent, highly selective FK506-binding protein 51 (FKBP51) inhibitor with a Ki of 6 nM and also enhances AKT2-AS160 binding.

SAFit2 is a highly potent, highly selective FK506-binding protein 51 (FKBP51) inhibitor with a Ki of 6 nM and also enhances AKT2-AS160 binding.

Specifications

Chemical Name or Material SAFit2
CAS 1643125-33-0
Color Pink
Purity Grade Notes Research
Molecular Formula C46H62N2O10
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (124.53 mM; Need ultrasonic)
SMILES O=C(N1[C@@H](CCCC1)C(O[C@@H](C2=CC(OCCN3CCOCC3)=CC=C2)CCC4=CC(OC)=C(OC)C=C4)=O)[C@@H](C5CCCCC5)C6=CC(OC)=C(OC)C(OC)=C6
Molecular Weight (g/mol) 802.99
Formula Weight 802.99
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.