missing translation for 'onlineSavingsMsg'
Learn More

Safflower oil, Food grade, MP Biomedicals™

Product Code. 15759150 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 15759150

Brand: MP Biomedicals 0210288890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 8001-23-8

Specifications

Color Yellow
Quantity 500 mL
Grade Food
Physical Form Oil
Chemical Name or Material Safflower oil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.