Learn More
MEDICOM Safewear PP Gown 25 g/m2, Snap
Product Code. 31781320 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31781320 Small Pack of 50
31781380 Medium Pack of 50
31781337 Large Pack of 50
31781303 X-Large Pack of 50
31781335 2X-Large Pack of 50
31781390 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31781320 Supplier MEDICOM Supplier No. 40.302/01
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
The polypropylene hygiene gown is comfortable, lightweight and breathable, designed to limit soiling in professional environments.
  • Disposable hygiene labcoats protect the work environment from particles coming from the operator's body (skin flakes, hair, sweat, etc.)
  • They are suitable for HACCP procedures
  • They help the operator protect themselves from the risk of stains related to their activity
Specifications
Size Small
Gender Unisex
Material Polypropylene
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance None
Style Knee-length
No. of Pockets None