Learn More
SCAT™ Safety Waste Caps V2.0 with Level Control
Product Code. 17853530 Shop All SCAT Europe Products
Change view
Click to view available options
Size:
B83
S55
S60/61
S65
S70/71
S90
Diameter (Metric) Outer:
2 x 2.3/3.2 mm
3 x 2.3 to 3.2 mm, 1 x 5 to 11.5 mm
3 x 2.3/3.2 mm
4 x 2.3/3.2 mm
4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm
Includes:
Electronic Level Control, Connector For Exhaust Filter
Electronic Level Control, Grounding Connection, Connector For Exhaust Filter
Mechanical Level Control, Connector For Exhaust Filter
Mechanical Level Control, Grounding Connection, Connector For Exhaust Filter
Unit Size:
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Diameter (Metric) Outer Includes unitSize
17853530 S60/61 3 x 2.3/3.2 mm Mechanical Level Control, Connector For Exhaust Filter Each
10450375 B83 4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm Electronic Level Control, Connector For Exhaust Filter Each
10231710 B83 4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm Mechanical Level Control, Connector For Exhaust Filter Each
10558130 S55 2 x 2.3/3.2 mm Electronic Level Control, Connector For Exhaust Filter Each
10889330 S55 3 x 2.3/3.2 mm Electronic Level Control, Connector For Exhaust Filter Each
10387969 S55 3 x 2.3/3.2 mm Mechanical Level Control, Connector For Exhaust Filter Each
10601324 S55 2 x 2.3/3.2 mm Mechanical Level Control, Connector For Exhaust Filter Each
16656214 S60/61 2 x 2.3/3.2 mm Electronic Level Control, Connector For Exhaust Filter Each
17049703 S60/61 3 x 2.3/3.2 mm Electronic Level Control, Connector For Exhaust Filter Each
17029703 S60/61 3 x 2.3/3.2 mm Electronic Level Control, Grounding Connection, Connector For Exhaust Filter Each
17039703 S60/61 4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm Electronic Level Control, Grounding Connection, Connector For Exhaust Filter Each
16646214 S60/61 2 x 2.3/3.2 mm Mechanical Level Control, Connector For Exhaust Filter Each
16666214 S60/61 2 x 2.3/3.2 mm Mechanical Level Control, Grounding Connection, Connector For Exhaust Filter Each
17019703 S60/61 3 x 2.3/3.2 mm Mechanical Level Control, Grounding Connection, Connector For Exhaust Filter Each
10394890 S65 4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm Mechanical Level Control, Connector For Exhaust Filter Each
10252850 S90 4 x 2.3/3.2 mm Electronic Level Control, Connector For Exhaust Filter Each
10881161 S90 4 x 2.3/3.2 mm Mechanical Level Control, Connector For Exhaust Filter Each
10324513 S90 4 x 2.3/3.2 mm, 1 x 5 to 11.5 mm Mechanical Level Control, Connector For Exhaust Filter Each
10849160 S70/71 3 x 2.3 to 3.2 mm, 1 x 5 to 11.5 mm Mechanical Level Control, Connector For Exhaust Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 17853530 Supplier SCAT™ Supplier No. 307961
  • ONLINE EXCLUSIVE PRICE
    £233.00
    £209.50 / Pack of 1
    Save £23.50
    10% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Safety Waste Caps with level control for additional automatic or mechanical monitoring of disposal container fill levels.
SCAT™ Safety Waste Caps provide maximum safety for your HPLC liquid disposal. Safely dispose of your liquid laboratory waste without contaminating the ambient air by using these Safety Waste Caps with integrated exhaust air filters. These high-performance filters regulate the air pressure balance and prevent the escape of vapors due to resulting overpressure in the disposal container.
  • High quality materials ensure maximum safety for lab and staff
  • Can be individually equipped with level indicators, safety funnels or grounding connections
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Connection port for a SCAT™ exhaust filter V3.0 on each Safety Waste Cap
  • Variety of closures, connections and thread sizes allows optimal adaptation to each HPLC situation
  • Versions with grounding connections, level controls or safety funnels offer for safe disposal
Specifications
For Use With (Equipment) Waste Disposal Container
Includes Mechanical Level Control, Connector For Exhaust Filter
Size S60/61
Type Safety Waste Cap
Diameter (Metric) Outer 3 x 2.3/3.2 mm