Learn More
SymLine™ Safety Waste Caps
Product Code. 17145501 Shop All SCAT Europe Products
Change view
Click to view available options
Thread Size:
GL45
S55
S60/61
S90
Includes:
1 GL25 (m), Filling Lance, Connector for Exhaust Filter
1 NPT 3/8 in. (f), Connection for Exhaust Filter
2 PFA-fittings, Tube Connector
2 PFA-fittings, Tube Connector, Collector for HPLC Waste
3 PFA-fittings, Collector for HPLC Waste
5 PFA-fittings, Collector for HPLC Waste
Level Control w/ Shut-off, Connector for Exhaust Filter
Level Control, Cable Connection, Connector for Exhaust Filter
Level Control, Connector for Exhaust Filter
Level Control, Filling Lance, Connector for Exhaust Filter
Unit Size:
1 sheet
Each
Product Code. Thread Size Includes unitSize
17145501 GL45 2 PFA-fittings, Tube Connector, Collector for HPLC Waste Each
17135501 GL45 3 PFA-fittings, Collector for HPLC Waste Each
16876011 GL45 5 PFA-fittings, Collector for HPLC Waste Each
15960386 S55 Level Control, Filling Lance, Connector for Exhaust Filter Each
15724922 S55 1 GL25 (m), Filling Lance, Connector for Exhaust Filter Each
17808743 S55 Level Control w/ Shut-off, Connector for Exhaust Filter Each
17079693 S60/61 Level Control, Cable Connection, Connector for Exhaust Filter Each
16866011 S60/61 2 PFA-fittings, Tube Connector Each
17813352 S60/61 Level Control w/ Shut-off, Connector for Exhaust Filter Each
17377409 S90 Level Control, Connector for Exhaust Filter Each
17838743 S90 Level Control w/ Shut-off, Connector for Exhaust Filter Each
18646478 S60/61 1 NPT 3/8 in. (f), Connection for Exhaust Filter 1 sheet
12 options
Product Code. 17145501
Brand: SymLine™ 306497
Only null left
Add to basket
Add to basket
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
  • Available with different thread sizes for attachment to pipe system
  • Different numbers of hose connections for disposal of liquid waste
  • Available with electronic level sensors that indicate the degree of filling of the disposal container
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
For Use With (Equipment) SymLine System for Capillaries with (OD) 1.6/2.3/3.2 mm
Includes 2 PFA-fittings, Tube Connector, Collector for HPLC Waste
Thread Size GL45
For Use With (Application) Waste Disposal
Type Safety Waste Cap