Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
| For Use With (Equipment) | SymLine System |
| Includes | Level Control, Cable Connection, Connector for Exhaust Filter |
| Thread Size | S60/61 |
| For Use With (Application) | Waste Disposal |
| Type | Safety Waste Cap |