Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
| For Use With (Equipment) | SymLine System for Capillaries with (OD) 1.6/2.3/3.2 mm |
| Includes | 2 PFA-fittings, Tube Connector |
| Thread Size | S60/61 |
| For Use With (Application) | Waste Disposal |
| Type | Safety Waste Cap |