Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Extension Funnel
Product Code. 17376429 Shop All SCAT Europe Products
Click to view available options
Type:
Electrically Conductive
Not Electrically Conductive
Includes:
Extension Funnel
Extension Funnel with Earthing Cable
Unit Size:
Each
Product Code. 17376429
Brand: SCAT 450120
Only null left
Add to basket
Add to basket
Safety Waste Cap 'LISA', V3.0 Extension Funnel is an accessory for LISA HPLC/UHPLC systems.
  • Accessory for Safety Waste Cap 'LISA,' V3.0
  • Extension Funnel with Hinged Lid
  • Made of PTFE for optimal chemical resistance
Specifications
For Use With (Equipment) HPLC / UHPLC Systems
Includes Extension Funnel with Earthing Cable
Material PTFE
Thread Size M30x35
Type Electrically Conductive