Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug
Product Code. 17326429 Shop All SCAT Europe Products
Change view
Click to view available options
Type:
Electrically Conductive
Not Electrically Conductive
Thread Size:
GL14
M30x35
NPT 1/8 in.
Includes:
Blind plug
Satellite Blind Plug
Unit Size:
Each
Product Code. Type Thread Size Includes unitSize
17326429 Electrically Conductive NPT 1/8 in. Blind plug Each
17316429 Electrically Conductive GL14 Blind plug Each
17336429 Not Electrically Conductive M30x35 Satellite Blind Plug Each
17366429 Electrically Conductive M30x35 Satellite Blind Plug Each
4 options
Product Code. 17326429
Brand: SCAT 160523
Only null left
Add to basket
Add to basket
Safety Waste Cap 'LISA', V3.0 Blind Plug is an accessory suitable for safely closing unused threaded holes in devices for the safe disposal of liquid HPLC/UHPLC waste.
  • Accessory for Safety Waste Cap 'LISA,' V3.0
  • For extending and attaching various components such as a funnel
  • Made of PTFE for optimal chemical resistance
Specifications
Type Electrically Conductive
Includes Blind plug
Thread Size NPT 1/8 in.
For Use With HPLC / UHPLC Systems
Material PTFE