Description
- Accessory for Safety Waste Cap 'LISA,' V3.0
- For extending and attaching various components such as a funnel
- Made of PTFE for optimal chemical resistance
Specifications
Specifications
| Type | Electrically Conductive |
| Includes | Blind plug |
| Thread Size | GL14 |
| For Use With | HPLC / UHPLC Systems |
| Material | PTFE |